Learn More
3-(1-Nitroethyl)-2-benzofuran-1(3H)-one, TRC
Product Code. 30467535
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467535 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30467535 Supplier Toronto Research Chemicals Supplier No. TRCN49589850MG
 Restricted Item
Spedizione stimata: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Chemical Name or Material 3-(1-Nitroethyl)-2-benzofuran-1(3H)-one
CAS 79017-08-6
Molecular Formula C10 H9 N O4
Quantity 50 mg
Synonym 3-(1-Nitroethyl)-3H-2-benzofuran-1-one, 3-(1-Nitroethyl)-1(3H)-isobenzofuranone
InChI Formula InChI=1S/C10H9NO4/c1-6(11(13)14)9-7-4-2-3-5-8(7)10(12)15-9/h2-6,9H,1H3
SMILES CC(C1OC(=O)c2ccccc12)[N+](=O)[O-]
IUPAC Name 3-(1-nitroethyl)-3H-2-benzofuran-1-one
Molecular Weight (g/mol) 207.2
Formula Weight 207.0532