missing translation for 'onlineSavingsMsg'
Learn More

3-(2,4-Dimethoxyphenyl)-1H-pyrazole, 98%, Thermo Scientific™

Catalog Number p-7033512
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
150433-17-3
C11H12N2O2
204.23
MFCD02091529
HEQGATYGFKDZIA-UHFFFAOYSA-N
3-2,4-dimethoxyphenyl-1h-pyrazole, 5-2,4-dimethoxyphenyl-1h-pyrazole, 3-2,4-dimethoxyphenyl pyrazole, 2,4-dimethoxy-1-pyrazol-3-ylbenzene, acmc-20anm3, 1h-pyrazole,3-2,4-dimethoxyphenyl, 3-2,4-dimethoxy-phenyl-1h-pyrazole
15859649
5-(2,4-dimethoxyphenyl)-1H-pyrazole
COC1=CC(OC)=C(C=C1)C1=CC=NN1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
150433-17-3
C11H12N2O2
204.23
MFCD02091529
HEQGATYGFKDZIA-UHFFFAOYSA-N
3-2,4-dimethoxyphenyl-1h-pyrazole, 5-2,4-dimethoxyphenyl-1h-pyrazole, 3-2,4-dimethoxyphenyl pyrazole, 2,4-dimethoxy-1-pyrazol-3-ylbenzene, acmc-20anm3, 1h-pyrazole,3-2,4-dimethoxyphenyl, 3-2,4-dimethoxy-phenyl-1h-pyrazole
15859649
5-(2,4-dimethoxyphenyl)-1H-pyrazole
COC1=CC(OC)=C(C=C1)C1=CC=NN1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-(2,4-Dimethoxyphenyl)-1H-pyrazole
Quantità 1 g
Peso formulazione 204.23
Percent Purity 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'