missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Carboxyvinyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number p-7046801
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
216144-91-1
C9H9BO4
191.98
MFCD01075738
QCHIEOGZUMAQKI-SNAWJCMRSA-N
3-2-carboxyvinyl benzeneboronic acid, e-3-3-boronophenyl acrylic acid, 3-boronocinnamic acid, 3-3-boronophenyl acrylic acid, 3-trans-2-carboxyvinyl phenylboronic acid, 3-2-carboxyvinyl phenylboronic acid, 3-phenylpropylester boronic acid, 3--e-2-carboxyvinyl benzeneboronic acid, 2e-3-3-dihydroxyboranyl phenyl prop-2-enoic acid, 3-3-dihydroxyboranyl phenyl prop-2-enoic acid
5844876
(E)-3-(3-boronophenyl)prop-2-enoic acid
OB(O)C1=CC=CC(\C=C\C(O)=O)=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
216144-91-1
C9H9BO4
191.98
MFCD01075738
QCHIEOGZUMAQKI-SNAWJCMRSA-N
3-2-carboxyvinyl benzeneboronic acid, e-3-3-boronophenyl acrylic acid, 3-boronocinnamic acid, 3-3-boronophenyl acrylic acid, 3-trans-2-carboxyvinyl phenylboronic acid, 3-2-carboxyvinyl phenylboronic acid, 3-phenylpropylester boronic acid, 3--e-2-carboxyvinyl benzeneboronic acid, 2e-3-3-dihydroxyboranyl phenyl prop-2-enoic acid, 3-3-dihydroxyboranyl phenyl prop-2-enoic acid
5844876
(E)-3-(3-boronophenyl)prop-2-enoic acid
OB(O)C1=CC=CC(\C=C\C(O)=O)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-(2-Carboxyvinyl)benzeneboronic acid
Punto di fusione ∼218°C
Quantità 250 mg
Peso formulazione 191.98
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'