Learn More
Sartorius™ 3.2 Standard Flexboy™ with TPE tubes
Marca: Sartorius™ FFB110911
Dettagli aggiuntivi : Peso : 21.00000kg
Specifica
10 L | |
Bags are designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products | |
Gamma Irradiated |
ISO11137 | |
3 Ports |