Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 1-Way Vacuum Stopcocks
Specifications
Specifications
| Tipo | 1-way Vacuum Stopcock |
| Foro spina | 4 mm |
| Materiale | Borosilicate Glass |
| Certificazione/Conformità | DIN 12545 |
| Include | Right-angled Sidearm, Hollow Glass Plug |