missing translation for 'onlineSavingsMsg'
Learn More

[3-(3-Methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol, 97%, Thermo Scientific™

Catalog Number p-3541101
Click to view available options
Quantità:
250 mg
1 g
Imballaggio:
Amber Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916766-84-2
C10H10N2O2
190.202
MFCD09817471
ZBIYTANHMLGZFB-UHFFFAOYSA-N
3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanol, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl alcohol, 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methan-1-ol, benzenemethanol,3-3-methyl-1,2,4-oxadiazol-5-yl
24229498
[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol
CC1=NOC(=N1)C2=CC(=CC=C2)CO
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916766-84-2
C10H10N2O2
190.202
MFCD09817471
ZBIYTANHMLGZFB-UHFFFAOYSA-N
3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanol, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl alcohol, 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methan-1-ol, benzenemethanol,3-3-methyl-1,2,4-oxadiazol-5-yl
24229498
[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol
CC1=NOC(=N1)C2=CC(=CC=C2)CO
Materiale o nome chimico [3-(3-Methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methanol
CAS min. % 96.5
CAS max % 100.0
Intervallo percentuale saggio 97%
Imballaggio Amber Glass Bottle
Quantità 250 mg
Peso formulazione 190.2
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'