missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)-4-(4-ethylphenyl)-5-methyl-4H-1,2,4-triazole, 97%, Thermo Scientific™

Catalog Number p-7040547
Click to view available options
Quantità:
100 mg
500 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1421263-53-7
C17H16BrN3
342.24
MFCD22683119
FSFAVMUCBYVKSQ-UHFFFAOYSA-N
3-4-bromophenyl-4-4-ethylphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-ethylphenyl-5-methyl-4h-1,2,4-triazole
97030404
3-(4-bromophenyl)-4-(4-ethylphenyl)-5-methyl-1,2,4-triazole
CCC1=CC=C(C=C1)N2C(=NN=C2C3=CC=C(C=C3)Br)C
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1421263-53-7
C17H16BrN3
342.24
MFCD22683119
FSFAVMUCBYVKSQ-UHFFFAOYSA-N
3-4-bromophenyl-4-4-ethylphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-ethylphenyl-5-methyl-4h-1,2,4-triazole
97030404
3-(4-bromophenyl)-4-(4-ethylphenyl)-5-methyl-1,2,4-triazole
CCC1=CC=C(C=C1)N2C(=NN=C2C3=CC=C(C=C3)Br)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-(4-Bromophenyl)-4-(4-ethylphenyl)-5-methyl-4H-1,2,4-triazole
Quantità 100 mg
Peso formulazione 342.24
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'