Learn More
3,4-Diaminopyridine, 97%
Descrizione
It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a potassium channel blocker in nerve terminals. It inhibits potassium channel efflux, increasing the duration of the action potential, which results in an increase in the duration of calcium channel opening and enhanced acetylcholine (ACh) release. Increased ACh availability at the motor end plate allows muscles to contract.
Solubility
Soluble in water (24 mg/ml at 20°C), alcohol. Slightly soluble in ether.
Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
Specifica
Specifica
| Materiale o nome chimico | 3,4-Diaminopyridine |
| Punto di fusione | 217°C to 220°C |
| Punto d'infiammabilità | 240°C (464°F) |
| Odore | Unpleasant |
| Quantità | 25 g |
| Numero UN | UN2811 |
| Beilstein | 110232 |
| Indice di Merck | 14,2986 |
| Informazioni di solubilità | Soluble in water (24mg/ml at 20°C),alcohol. Slightly soluble in ether. |
| Peso formulazione | 109.13 |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.