missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methylphenyl)-1H-pyrazole-4-carboxaldehyde, 97%, Thermo Scientific™

Catalog Number 11395499
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
350988-62-4
C11H10N2O
186.21
MFCD01133694
CKEZOUSRIZFLQE-UHFFFAOYSA-N
3-4-methylphenyl-1h-pyrazole-4-carbaldehyde, 5-4-methylphenyl-1h-pyrazole-4-carbaldehyde, 3-p-tolyl-1h-pyrazole-4-carbaldehyde, 3-p-tolyl-1h-pyrazole-4-carboxaldehyde, 5-p-tolyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-4-methylphenyl, 3-4-methylphenyl pyrazole-4-carbaldehyde, 3-4-methylphenyl-2h-pyrazole-4-carbaldehyde, methylphenylpyrazolecarbaldehyde
2760667
5-(4-methylphenyl)-1H-pyrazole-4-carbaldehyde
CC1=CC=C(C=C1)C1=C(C=O)C=NN1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
350988-62-4
C11H10N2O
186.21
MFCD01133694
CKEZOUSRIZFLQE-UHFFFAOYSA-N
3-4-methylphenyl-1h-pyrazole-4-carbaldehyde, 5-4-methylphenyl-1h-pyrazole-4-carbaldehyde, 3-p-tolyl-1h-pyrazole-4-carbaldehyde, 3-p-tolyl-1h-pyrazole-4-carboxaldehyde, 5-p-tolyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3-4-methylphenyl, 3-4-methylphenyl pyrazole-4-carbaldehyde, 3-4-methylphenyl-2h-pyrazole-4-carbaldehyde, methylphenylpyrazolecarbaldehyde
2760667
5-(4-methylphenyl)-1H-pyrazole-4-carbaldehyde
CC1=CC=C(C=C1)C1=C(C=O)C=NN1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-(4-Methylphenyl)-1H-pyrazole-4-carboxaldehyde
Quantità 1 g
Sensibilità Air Sensitive
Peso formulazione 186.22
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'