missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl bromide, 98%, Thermo Scientific™

Catalog Number p-7024613
Click to view available options
Quantità:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
32247-96-4
C9H5BrF6
307.03
MFCD00009905
ATLQGZVLWOURFU-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
122573
1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
32247-96-4
C9H5BrF6
307.03
MFCD00009905
ATLQGZVLWOURFU-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
122573
1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Bis(trifluoromethyl)benzyl bromide is used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry in enantioselective synthesis of non-peptidic neurokinin NK1 receptor antagonist.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3,5-Bis(trifluoromethyl)benzyl bromide
Densità 1.675
Punti di ebollizione 136°C to 140°C (14 mmHg)
Punto d'infiammabilità >100°C (212°F)
Indice di rifrazione 1.445
Quantità 1 g
Numero UN UN3265
Beilstein 656506
Informazioni di solubilità Difficult to mix.
Peso formulazione 307.04
Percent Purity 98%
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'