Learn More
3-[5-Chloro-2-(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™
Product Code. 15477885
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477885 250 mg 250mg
15467885 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15477885 Supplier Thermo Scientific Alfa Aesar Supplier No. H31746.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 916420-80-9
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.62
MDL Number MFCD09025402
InChI Key PWUOREFPPQVKFH-UHFFFAOYSA-N
Synonym 3-5-chloro-2-trifluoromethyl phenyl propionic acid, 3-5-chloro-2-trifluoromethyl phenyl propanoic acid
PubChem CID 46737571
IUPAC Name 3-[5-chloro-2-(trifluoromethyl)phenyl]propanoic acid
SMILES OC(=O)CCC1=C(C=CC(Cl)=C1)C(F)(F)F
Specifications
Melting Point 42°C to 45°C
Quantity 250 mg
Formula Weight 252.62
Percent Purity 97%
Chemical Name or Material 3-[5-Chloro-2-(trifluoromethyl)phenyl]propionic acid