missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-methoxybenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number 15443527
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
105969-16-2
C8H9F2NO
173.163
MFCD04115915
MCQREBXBVNPGJZ-UHFFFAOYSA-N
3,5-difluoro-4-methoxybenzylamine, 3,5-difluoro-4-methoxyphenyl methanamine, 1-3,5-difluoro-4-methoxyphenyl methanamine, acmc-20ancx, 3,5-difluoro-4-methoxyphenyl methylamine, 3,5-difluoro-4-methoxy-phenyl methanamine, benzenemethanamine,3,5-difluoro-4-methoxy, 3,5-bis fluoranyl-4-methoxy-phenyl methanamine
14626469
(3,5-difluoro-4-methoxyphenyl)methanamine
COC1=C(C=C(C=C1F)CN)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
105969-16-2
C8H9F2NO
173.163
MFCD04115915
MCQREBXBVNPGJZ-UHFFFAOYSA-N
3,5-difluoro-4-methoxybenzylamine, 3,5-difluoro-4-methoxyphenyl methanamine, 1-3,5-difluoro-4-methoxyphenyl methanamine, acmc-20ancx, 3,5-difluoro-4-methoxyphenyl methylamine, 3,5-difluoro-4-methoxy-phenyl methanamine, benzenemethanamine,3,5-difluoro-4-methoxy, 3,5-bis fluoranyl-4-methoxy-phenyl methanamine
14626469
(3,5-difluoro-4-methoxyphenyl)methanamine
COC1=C(C=C(C=C1F)CN)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3,5-Difluoro-4-methoxybenzylamine
Indice di rifrazione 1.498
Quantità 250 mg
Numero UN UN2735
Sensibilità Air Sensitive
Peso formulazione 173.16
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'