Learn More
3',5'-Difluoroacetophenone, 97%
Product Code. 11449816
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449816 25 g 25g
11429816 1 g 1g
11439816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11449816 Supplier Thermo Scientific Alfa Aesar Supplier No. B20301.14
 Restricted Item
Spedizione stimata: 02-12-2026
Add to basket
CAS: 123577-99-1 | C8H6F2O | 156.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123577-99-1
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00042489
InChI Key OXJLDNSPGPBDCP-UHFFFAOYSA-N
Synonym 3',5'-difluoroacetophenone, 3,5-difluoroacetophenone, 1-3,5-difluorophenyl ethanone, 1-3,5-difluorophenyl ethan-1-one, ethanone, 1-3,5-difluorophenyl, 1-acetyl-3,5-difluorobenzene, 1-3,5-difluoro-phenyl-ethanone, pubchem2179, acmc-1cacd, 3,5-di-fluoroacetophenone
PubChem CID 518596
SMILES CC(=O)C1=CC(F)=CC(F)=C1
Specifications
Melting Point 37°C to 41°C
Boiling Point 94°C to 95°C (13 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.4876
Quantity 25 g
Beilstein 6798740
IUPAC Name 1-(3,5-difluorophenyl)ethan-1-one
Formula Weight 156.13
Percent Purity 97%
Chemical Name or Material 3',5'-Difluoroacetophenone