missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluorobenzeneboronic acid neopentyl glycol ester, 98+%

Catalog Number p-7058474
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
216393-57-6
C11H13BF2O2
226.03
MFCD01318139
OLHRAEPCGZMMTI-UHFFFAOYSA-N
2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 3,5-difluorobenzeneboronic acid neopentyl glycol ester, 3,5-difluorophenylboronic acid neopentyl glycol ester, 3,5-difluorobenzeneboronic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborane, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborine, 1,3,2-dioxaborinane, 2-3,5-difluorophenyl-5,5-dimethyl, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyldihydro-4h-1,3,2-dioxaborin
2774056
2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
B1(OCC(CO1)(C)C)C2=CC(=CC(=C2)F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
216393-57-6
C11H13BF2O2
226.03
MFCD01318139
OLHRAEPCGZMMTI-UHFFFAOYSA-N
2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborinane, 3,5-difluorobenzeneboronic acid neopentyl glycol ester, 3,5-difluorophenylboronic acid neopentyl glycol ester, 3,5-difluorobenzeneboronic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborane, 2-3,5-difluorophenyl-5,5-dimethyl-1,3,2-dioxaborine, 1,3,2-dioxaborinane, 2-3,5-difluorophenyl-5,5-dimethyl, 3,5-difluorobenzenebornoic acid neopentyl glycol cyclic ester, 2-3,5-difluorophenyl-5,5-dimethyldihydro-4h-1,3,2-dioxaborin
2774056
2-(3,5-difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
B1(OCC(CO1)(C)C)C2=CC(=CC(=C2)F)F
Materiale o nome chimico 3,5-Difluorobenzeneboronic acid neopentyl glycol ester
Punto di fusione 43°C to 45°C
Quantità 1 g
Peso formulazione 226.03
Percent Purity ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'