Learn More
Thermo Scientific Alfa Aesar 50ML 3,5-Dimethylphenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal^t bottl
Marca:  Thermo Scientific Alfa Aesar H58027.AD
50ML 3,5-Dimethylphenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal^t bottl
Dettagli aggiuntivi : CAS : 312692-98-1 Peso : 0.00050kg Transport : UN number : 3399 Chem class : 4.3 Pack group : II
Codice prodotto. 15485305