Learn More
3-Acetyl-2,5-dimethylfuran, 98%
Product Code. 11472977
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472977 250 g 250g
11452977 10 g 10g
11462977 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11472977 Supplier Thermo Scientific Alfa Aesar Supplier No. A15545.30
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 10599-70-9 | C8H10O2 | 138.166 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10599-70-9
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00010281
InChI Key KBSVBCHYXYXDAG-UHFFFAOYSA-N
Synonym 3-acetyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl ethanone, ethanone, 1-2,5-dimethyl-3-furanyl, 3-acetyl-2,5-dimethyl furan, 2,5-dimethyl-3-acetylfuran, 1-2,5-dimethyl-3-furyl ethan-1-one, 2,5-dimethyl-3-furyl methyl ketone, 1-2,5-dimethyl-3-furanyl ethanone, 1-2,5-dimethyl-3-furyl ethanone, unii-798v2t7zbv
PubChem CID 61527
IUPAC Name 1-(2,5-dimethylfuran-3-yl)ethanone
SMILES CC1=CC(=C(O1)C)C(=O)C
Specifications
Density 1.038
Boiling Point 198°C
Flash Point 78°C (172°F)
Refractive Index 1.4865
Quantity 250 g
Beilstein 112094
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 3-Acetyl-2,5-dimethylfuran