Learn More
3-Acetylacetanilide, TRC
Product Code. 30316208
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316208 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30316208 Supplier Toronto Research Chemicals Supplier No. TRCA133000100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Acetylacetanilide
CAS 7463-31-2
Molecular Formula C10H11NO2
Quantity 100 mg
Synonym N-(3-Acetylphenyl)acetamide, 2-Acetamidoacetophenone, NSC 404340
InChI Formula InChI=1S/C10H11NO2/c1-7(12)9-4-3-5-10(6-9)11-8(2)13/h3-6H,1-2H3,(H,11,13)
SMILES CC(=O)Nc1cccc(c1)C(=O)C
IUPAC Name N-(3-acetylphenyl)acetamide
Molecular Weight (g/mol) 177.2
Formula Weight 177.079