Learn More
3-Amino-1,2,4-triazole (Amitrole), TRC
Specifica
Specifica
| Materiale o nome chimico | Amitrole |
| CAS | 61-82-5 |
| Formula molecolare | C2 H4 N4 |
| Quantità | 10 g |
| Sinonimo | 1H-1,2,4-Triazol-3-amine (9CI), 1H-1,2,4-Triazole, 3-amino- (7CI), s-Triazole, 3-amino- (8CI), (4H-1,2,4-Triazol-3-yl)amine, 1,2,4-Triazol-5-amine, 1H-1,2,4-Triazol-3-ylamine, 1H-1,2,4-Triazolamine, 2,3,5,6-Tetraazabicyclo[2.1.1]hex-1-ene, 2-Amino-1,3,4-triazole, 3,A-T, 3-Amino-1,2,4-triazole, 3-Amino-1H-1,2,4-triazole, 3-Amino-2H-1,2,4-triazole, 3-Amino-s-triazole, 3-Aminotriazole, 4H-3-Amino-1,2,4-triazole, 5-Amino-1,2,4-triazole, 5-Amino-1H-1,2,4-triazole, A 0432, AT, ATA, ATA (amine), Amerol, Aminotriazole (plant regulator), Amitrol, Amitrol 90, Amitrol T, Amitrole, Amizol, Azaplant, Azolan, Cytrol, Cytrole, ENT 25445, Herbidal total, Herbizole, Maxata, NSC 34809, NSC 7243, TMG, Weedazol |
| InChI Formula | InChI=1S/C2H4N4/c3-2-4-1-5-6-2/h1H,(H3,3,4,5,6) |
| SMILES | Nc1nc[nH]n1 |
| IUPAC Name | 1H-1,2,4-triazol-3-amine |
| Molecular Weight (g/mol) | 84.08 |
| Peso formulazione | 84.0436 |