Learn More
3-Benzoyl-1,1,1-trifluoroacetone, 98+%
Product Code. 11438986
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438986 25 g 25g
11448986 100 g 100g
11458986 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11438986 Supplier Thermo Scientific Alfa Aesar Supplier No. A17110.14
 Restricted Item
In stock
Add to basket
CAS: 326-06-7 | C10H7F3O2 | 216.16 g/mol
3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.
Solubility
Sparingly Soluble in water (0.24 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 326-06-7
Molecular Formula C10H7F3O2
Molecular Weight (g/mol) 216.16
MDL Number MFCD00000425
InChI Key VVXLFFIFNVKFBD-UHFFFAOYSA-N
Synonym benzoyl-1,1,1-trifluoroacetone, benzoyltrifluoroacetone, 4,4,4-trifluoro-1-phenyl-1,3-butanedione, benzoyl trifluoroacetyl methane, 3-benzoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-phenyl, 4,4,4-trifluoro-1-phenyl-butane-1,3-dione, 1-benzoyl-3,3,3-trifluoroacetone, .omega.-trifluoroacetyl acetophenone, 1-benzoyl-3,3,3-trifluoro-2-propanone
PubChem CID 67589
IUPAC Name 4,4,4-trifluoro-1-phenylbutane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1
Specifications
Melting Point 37°C to 40°C
Density 1.113
Boiling Point 224°C
Flash Point 98°C (208°F)
Quantity 25 g
Beilstein 1875083
Solubility Information Sparingly Soluble in water (0.24g/L) (25°C).
Formula Weight 216.16
Percent Purity ≥98%
Chemical Name or Material 3-Benzoyl-1,1,1-trifluoroacetone
compliance-icons
Product Identifier
  • 3-Benzoyl-1,1,1-trifluoroacetone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Benzoyl-1,1,1-trifluoroacetone