Learn More
3-Bromo-4-pyridinecarboxylic acid, 97%, Thermo Scientific™
Product Code. 11946441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11946441 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11946441 Supplier Thermo Scientific Acros Supplier No. 442750010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 13959-02-9
Melting Point 241°C to 244°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H4BrNO2
MDL Number MFCD00040944
Quantity 1 g
Synonym 3-bromoisonicotinic acid, 3-bromo-4-pyridinecarboxylic acid, 3-bromo-isonicotinic acid, 4-pyridinecarboxylic acid, 3-bromo, 5-bromoisonicotinic acid, pubchem13576, 3-bromo isonicotinic acid, acmc-1cef2, ksc174i2l, 3-bromo-4-pyridine carboxylic acid
InChI Key AVXWWBFBRTXBRM-UHFFFAOYSA-N
SMILES OC(=O)C1=C(Br)C=NC=C1
IUPAC Name 3-bromopyridine-4-carboxylic acid
Molecular Weight (g/mol) 202.01
PubChem CID 817972
Formula Weight 202.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-pyridinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-pyridinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.