Learn More
3-Bromo-6-pyridinecarbonitrile, TRC
Product Code. 30334819
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334819 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30334819 Supplier Toronto Research Chemicals Supplier No. TRCB6867505G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Bromo-6-pyridinecarbonitrile
CAS 97483-77-7
Molecular Formula C6 H3 Br N2
Quantity 5 g
Synonym 2-Pyridinecarbonitrile, 5-bromo-, 5-Bromo-2-pyridinecarbonitrile, 2-Cyano-5-bromopyridine, 3-Bromo-6-cyanopyridine, 3-Bromo-6-pyridinecarbonitrile, 5-Bromo-2-cyanopyridine, 5-Bromopicolinonitrile, 5-Bromopyridine-2-carbonitrile
InChI Formula InChI=1S/C6H3BrN2/c7-5-1-2-6(3-8)9-4-5/h1-2,4H
SMILES Brc1ccc(nc1)C#N
IUPAC Name 5-bromopyridine-2-carbonitrile
Molecular Weight (g/mol) 183.01
Formula Weight 181.948