Learn More
3-Bromopentane, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromopentane |
| CAS | 1809-10-5 |
| Molecular Formula | C5H11Br |
| Quantity | 50 g |
| Synonym | 3-Pentyl Bromide |
| InChI Formula | InChI=1S/C5H11Br/c1-3-5(6)4-2/h5H,3-4H2,1-2H3 |
| SMILES | CCC(Br)CC |
| IUPAC Name | 3-bromopentane |
| Molecular Weight (g/mol) | 151.04 |
| Formula Weight | 150.0044 |