missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number 15443517
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
202001-00-1
C8H6ClFO2
188.582
MFCD03788475
GFLQMGYDUPLXHZ-UHFFFAOYSA-N
3-chloro-5-fluorophenylacetic acid, 2-3-chloro-5-fluorophenyl acetic acid, 5-chloro-3-fluorophenylacetic acid, 3-chloro-5-fluorophenyl acetic acid, benzeneacetic acid, 3-chloro-5-fluoro, 3-chloro-5-fluoro-phenyl-acetic acid, 2-5-chloro-3-fluorophenyl acetic acid, pubchem4266, 2-3-chloro-5-fluoro-phenyl acetic acid, acmc-1ckku
2734837
2-(3-chloro-5-fluorophenyl)acetic acid
C1=C(C=C(C=C1F)Cl)CC(=O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
202001-00-1
C8H6ClFO2
188.582
MFCD03788475
GFLQMGYDUPLXHZ-UHFFFAOYSA-N
3-chloro-5-fluorophenylacetic acid, 2-3-chloro-5-fluorophenyl acetic acid, 5-chloro-3-fluorophenylacetic acid, 3-chloro-5-fluorophenyl acetic acid, benzeneacetic acid, 3-chloro-5-fluoro, 3-chloro-5-fluoro-phenyl-acetic acid, 2-5-chloro-3-fluorophenyl acetic acid, pubchem4266, 2-3-chloro-5-fluoro-phenyl acetic acid, acmc-1ckku
2734837
2-(3-chloro-5-fluorophenyl)acetic acid
C1=C(C=C(C=C1F)Cl)CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Chloro-5-fluorophenylacetic acid
Punto di fusione 82°C to 85°C
Quantità 250 mg
Peso formulazione 188.59
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'