Learn More
3-Chloroiminodibenzyl, TRC
Product Code. 30367903
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367903 250 g -
30358842 5 g 5g
30357219 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30367903 Supplier Toronto Research Chemicals Supplier No. TRCC366920250G
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32943-25-2
Molecular Formula C14 H12 Cl N
Molecular Weight (g/mol) 229.7
Synonym 3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine, Clomipramine Hydrochloride Imp. F (EP)
IUPAC Name 2-chloro-6,11-dihydro-5H-benzo[b][1]benzazepine
SMILES Clc1ccc2CCc3ccccc3Nc2c1
Specifications
Quantity 250 g
InChI Formula InChI=1S/C14H12ClN/c15-12-8-7-11-6-5-10-3-1-2-4-13(10)16-14(11)9-12/h1-4,7-9,16H,5-6H2
Formula Weight 229.0658
Chemical Name or Material 3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine