missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number 15442379
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
214360-46-0
C13H16BNO2
229.09
MFCD03789262
FIGQEPXOSAFKTA-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyanophenylboronic acid pinacol ester, 3-cyanophenylboronic acid, pinacol ester, benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-cyanobenzeneboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, pubchem15603, 3-cyanophenylboronicacidpinacolester, 3-cyanobenzeneboronic acid, pinacol ester
2760590
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C#N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
214360-46-0
C13H16BNO2
229.09
MFCD03789262
FIGQEPXOSAFKTA-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyanophenylboronic acid pinacol ester, 3-cyanophenylboronic acid, pinacol ester, benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-cyanobenzeneboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, pubchem15603, 3-cyanophenylboronicacidpinacolester, 3-cyanobenzeneboronic acid, pinacol ester
2760590
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Cyanobenzeneboronic acid pinacol ester
Punto di fusione 78°C to 82°C (decomposition)
Colore White
Quantità 5 g
Peso formulazione 229.09
Percent Purity 97%
Forma fisica Solid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'