Learn More
3-(Dimethylamino)propyl-2-(benzylamino)benzoate, Mikromol
Product Code. 30459557
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30459557 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459557 Supplier Mikromol Supplier No. MM0142.020100
 Restricted Item
Spedizione stimata: 30-11-2026
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(Dimethylamino)propyl-2-(benzylamino)benzoate, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Dimethylaminopropyl-2-benzylaminobenzoate
CAS 87453-76-7
Analyte or Component Names 3-Dimethylaminopropyl-2-benzylaminobenzoate
Shipping Condition Room Temperature
Molecular Formula C19 H24 N2 O2
Certifications/Compliance ISO 9001
Synonym 2-[(Phenylmethyl)amino]benzoic acid 3-(Dimethylamino)propyl ester
InChI Formula InChI=1S/C19H24N2O2/c1-21(2)13-8-14-23-19(22)17-11-6-7-12-18(17)20-15-16-9-4-3-5-10-16/h3-7,9-12,20H,8,13-15H2,1-2H3
Solution Type Neat
SMILES CN(C)CCCOC(=O)c1ccccc1NCc2ccccc2
IUPAC Name 3-(dimethylamino)propyl 2-(benzylamino)benzoate
Molecular Weight (g/mol) 312.41
Quantity 100 mg
Formula Weight 312.1838 g/mol