Learn More
Alfa Aesar™ 3-Dimethylaminopropyl chloride hydrochloride, 98%
Marca: Alfa Aesar™ A15459.0E
Dettagli aggiuntivi : CAS : 5407-04-5 Peso : 2.50000kg
Quantità | 2500g |
---|
Identificatori chimici
5407-04-5 | |
158.07 | |
LJQNMDZRCXJETK-UHFFFAOYSA-N | |
94308 | |
CN(C)CCCCl.Cl |
C5H13Cl2N | |
MFCD00012521 | |
3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
3-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
Specifica
3-Dimethylaminopropyl chloride hydrochloride | |
138°C to 144°C | |
5407-04-5 | |
MFCD00012521 | |
3-dimethylaminopropylchloride hydrochloride, 3-chloro-n,n-dimethylpropan-1-amine hydrochloride, 3-dimethylaminopropyl chloride hydrochloride, 3-dimethylamino propyl chloride hydrochloride, n,n-dimethyl-3-chloropropylamine hydrochloride, 3-chloropropyl dimethylamine hydrochloride, 3-chloro-n,n-dimethylpropylamine hydrochloride, dimethylaminopropyl chloride hydrochloride, ccris 7054, dimethylaminopropyl chloride, hydrochloride | |
CN(C)CCCCl.Cl | |
158.07 | |
158.07 |
Hygroscopic | |
2500g | |
C5H13Cl2N | |
3670046 | |
LJQNMDZRCXJETK-UHFFFAOYSA-N | |
3-chloro-N,N-dimethylpropan-1-amine;hydrochloride | |
94308 | |
98% |