missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methoxybenzylamine, 97%, Thermo Scientific™

Catalog Number p-7031280
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916420-49-0
C8H10FNO
155.172
MFCD09025368
FVGJLRXKGHPGOW-UHFFFAOYSA-N
3-fluoro-2-methoxybenzylamine, 3-fluoro-2-methoxyphenyl methanamine, 1-3-fluoro-2-methoxyphenyl methanamine, 3-fluoro-2-methoxyphenyl methylamine
46737535
(3-fluoro-2-methoxyphenyl)methanamine
COC1=C(C=CC=C1F)CN
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916420-49-0
C8H10FNO
155.172
MFCD09025368
FVGJLRXKGHPGOW-UHFFFAOYSA-N
3-fluoro-2-methoxybenzylamine, 3-fluoro-2-methoxyphenyl methanamine, 1-3-fluoro-2-methoxyphenyl methanamine, 3-fluoro-2-methoxyphenyl methylamine
46737535
(3-fluoro-2-methoxyphenyl)methanamine
COC1=C(C=CC=C1F)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Fluoro-2-methoxybenzylamine
Indice di rifrazione 1.5142
Quantità 250 mg
Numero UN UN2735
Sensibilità Air Sensitive
Peso formulazione 155.17
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'