Learn More
3-Hydroxy-2-propyl-4-pentenoic Acid Ethyl Ester(Mixture of diastereomers), TRC
Specifica
Specifica
| Materiale o nome chimico | 3-Hydroxy-2-propyl-4-pentenoic Acid Ethyl Ester(Mixture of diastereomers) |
| CAS | 96107-27-6 |
| Formula molecolare | C10H18O3 |
| Quantità | 100 mg |
| Sinonimo | Ethyl 2-Propyl-3-hydroxy-4-pentenoate |
| InChI Formula | InChI=1S/C10H18O3/c1-4-7-8(9(11)5-2)10(12)13-6-3/h5,8-9,11H,2,4,6-7H2,1,3H3 |
| SMILES | CCCC(C(O)C=C)C(=O)OCC |
| IUPAC Name | ethyl 3-hydroxy-2-propylpent-4-enoate |
| Molecular Weight (g/mol) | 186.25 |
| Peso formulazione | 186.1256 |