missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-5-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog Number 15445407
Click to view available options
Quantità:
250 mg
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
42732-49-0
C6H7NO
109.128
MFCD00661297
RYJNCIGFPWGVPA-UHFFFAOYSA-N
3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg
224753
5-methylpyridin-3-ol
CC1=CC(=CN=C1)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
42732-49-0
C6H7NO
109.128
MFCD00661297
RYJNCIGFPWGVPA-UHFFFAOYSA-N
3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg
224753
5-methylpyridin-3-ol
CC1=CC(=CN=C1)O

3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Hydroxy-5-methylpyridine
Punto di fusione 136°C to 140°C
Quantità 250 mg
Informazioni di solubilità Slightly soluble in water.
Peso formulazione 109.13
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'