missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Hydroxymethyl-4-methoxybenzeneboronic acid, 98%

Marca:  Alfa Aesar™ H64738.06

Dettagli aggiuntivi : CAS : 908142-03-0 Peso : 0.00500kg

Quantità 5g

 vedi altre versioni di questo prodotto

Codice prodotto. 15442049

  • 262.00€ / 5g


Spedizione stimata: 25-06-2021
Aggiungi al carrello

Descrizione

Descrizione

Applications
It is an important intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifica

Identificatori chimici

908142-03-0
181.982
DOQUCBSZQOPLGT-UHFFFAOYSA-N
53407782
B(C1=CC(=C(C=C1)OC)CO)(O)O
C8H11BO4
MFCD14155851
3-hydroxymethyl-4-methoxyphenyl boronic acid, 3-hydroxymethyl-4-methoxyphenylboronic acid, ksc005q4b, 3-hydroxymethyl-4-methoxybenzeneboronic acid, 3-hydroxymethyl-4-methoxyphenyl boronicacid, b-3-hydroxymethyl-4-methoxyphenyl-boronic acid, boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
[3-(hydroxymethyl)-4-methoxyphenyl]boronic acid

Specifica

3-Hydroxymethyl-4-methoxybenzeneboronic acid
5g
C8H11BO4
15501236
Slightly soluble in water.
B(C1=CC(=C(C=C1)OC)CO)(O)O
181.982
181.98
98%
White
908142-03-0
MFCD14155851
3-hydroxymethyl-4-methoxyphenyl boronic acid, 3-hydroxymethyl-4-methoxyphenylboronic acid, ksc005q4b, 3-hydroxymethyl-4-methoxybenzeneboronic acid, 3-hydroxymethyl-4-methoxyphenyl boronicacid, b-3-hydroxymethyl-4-methoxyphenyl-boronic acid, boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
DOQUCBSZQOPLGT-UHFFFAOYSA-N
[3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
53407782
Powder
Videos
SDS Documenti
Certificati