missing translation for 'onlineSavingsMsg'
Learn More

3-(Hydroxymethyl)Benzamide (~85%), TRC

Codice prodotto. 30405099
Cambia vista
Click to view available options
Quantità:
25 mg
50 mg
250 mg
Dimensione della confezione:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30405099 250 mg 250mg
30394088 25 mg -
30402625 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30405099 Fornitore Toronto Research Chemicals N. del fornitore TRCH126920250MG

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 126926-34-9
Formula molecolare C8H9NO2
Molecular Weight (g/mol) 151.16
Sinonimo 3-(Hydroxymethyl)benzamide
IUPAC Name 3-(hydroxymethyl)benzamide
SMILES NC(C1=CC=CC(CO)=C1)=O

Specifications

Materiale o nome chimico 3-(Hydroxymethyl)Benzamide
Quantità 250 mg
InChI Formula InChI=1S/C8H9NO2/c9-8(11)7-3-1-2-6(4-7)5-10/h1-4,10H,5H2,(H2,9,11)
Peso formulazione 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.