missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-7031577
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916420-92-3
C9H6ClF3O2
238.59
MFCD09025410
IHMFMHOCMZSKNI-UHFFFAOYSA-N
3-methoxy-5-trifluoromethyl benzoyl chloride, 5-methoxy-3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-methoxy-5-trifluoromethyl
46737576
3-methoxy-5-(trifluoromethyl)benzoyl chloride
COC1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916420-92-3
C9H6ClF3O2
238.59
MFCD09025410
IHMFMHOCMZSKNI-UHFFFAOYSA-N
3-methoxy-5-trifluoromethyl benzoyl chloride, 5-methoxy-3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-methoxy-5-trifluoromethyl
46737576
3-methoxy-5-(trifluoromethyl)benzoyl chloride
COC1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Methoxy-5-(trifluoromethyl)benzoyl chloride
Indice di rifrazione 1.4915
Quantità 1 g
Numero UN UN3265
Sensibilità Moisture sensitive
Informazioni di solubilità Not miscible or difficult to mix with water.
Peso formulazione 238.59
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'