Learn More
3-Methoxyquinoline-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Codice prodotto. 15440906
Cambia vista
Click to view available options
Quantità:
250 mg
1 g
Dimensione della confezione:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15440906 1 g 1g
15450906 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15440906 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H54211.03
 Articolo soggetto a restrizioni
Spedizione stimata: 13-11-2026
Aggiungi al carrello
CAS: 881656-35-5 | Molecular Formula: C16H20BNO3 | Molecular Weight: 285.15 g/mol
3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxyquinoline-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 881656-35-5
Formula molecolare C16H20BNO3
Molecular Weight (g/mol) 285.15
Numero MDL MFCD20527212
InChI Key MGEKTRDPBDREKY-UHFFFAOYSA-N
Sinonimo 3-methoxyquinoline-5-boronic acid pinacol ester, 3-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-quinoline, 3-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 3-methoxy-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl-quinoline
PubChem CID 68783972
IUPAC Name 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES COC1=CN=C2C=CC=C(B3OC(C)(C)C(C)(C)O3)C2=C1
Specifica
Materiale o nome chimico 3-Methoxyquinoline-5-boronic acid pinacol ester
Quantità 1 g
Peso formulazione 285.15
Percent Purity 96%