missing translation for 'onlineSavingsMsg'
Learn More

3-Methylamino-1-propanol, 95%, Thermo Scientific Chemicals

Catalog Number p-7039536
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
42055-15-2
C4H11NO
89.138
MFCD06797456
KRGXWTOLFOPIKV-UHFFFAOYSA-N
3-methylamino propan-1-ol, 3-methylamino-1-propanol, 3-methylaminopropanol, 3-methylamino-propanol, 3-methylamino propanol, acmc-1argo, 3-methylamino-propan-1-ol, 3-hydroxypropyl methylamine, n-methyl-3-hydroxypropylamine
10148986
3-(methylamino)propan-1-ol
CNCCCO
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
42055-15-2
C4H11NO
89.138
MFCD06797456
KRGXWTOLFOPIKV-UHFFFAOYSA-N
3-methylamino propan-1-ol, 3-methylamino-1-propanol, 3-methylaminopropanol, 3-methylamino-propanol, 3-methylamino propanol, acmc-1argo, 3-methylamino-propan-1-ol, 3-hydroxypropyl methylamine, n-methyl-3-hydroxypropylamine
10148986
3-(methylamino)propan-1-ol
CNCCCO

3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylamino-1-propanol is an alkanolamine that is used to redisperse compacted solids (such as antiperspirant materials) in order to analyze its components.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Methylamino-1-propanol
Densità 0.98
Punti di ebollizione 169°C
Punto d'infiammabilità 82°C (180°F)
Formula lineare CH3NH(CH2)3OH
Indice di rifrazione 1.44
Quantità 1 g
Numero UN UN2735
Informazioni di solubilità Slightly soluble in water.
Peso formulazione 89.14
Percent Purity 95%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'