Learn More
3-Methylbenzophenone, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantità:
10 g
50 g
Descrizione
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifica
Materiale o nome chimico | 3-Methylbenzophenone |
Densità | 1.093 |
Punti di ebollizione | 316°C |
Punto d'infiammabilità | >110°C (230°F) |
Indice di rifrazione | 1.598 |
Quantità | 10 g |
Beilstein | 2045488 |
Informazioni di solubilità | Difficult to mix. |
Peso formulazione | 196.25 |
Percent Purity | 99% |