Learn More
3-Methylcrotonic Acid, TRC
Specifica
Specifica
| Materiale o nome chimico | 3-Methyl-2-butenoic Acid |
| CAS | 541-47-9 |
| Formula molecolare | C5 H8 O2 |
| Quantità | 100 g |
| Sinonimo | 2-Butenoic acid, 3-methyl-, Crotonic acid, 3-methyl- (8CI), 3-Methyl-2-butenoic acid, 3,3-Dimethylacrylic acid, 3-Methylcrotonic acid, NSC 2549, NSC 97179, Senecioic acid, β,β-Dimethylacrylic acid, β-Methylcrotonic acid |
| InChI Formula | InChI=1S/C5H8O2/c1-4(2)3-5(6)7/h3H,1-2H3,(H,6,7) |
| SMILES | CC(=CC(=O)O)C |
| IUPAC Name | 3-methylbut-2-enoic acid |
| Molecular Weight (g/mol) | 100.116 |
| Peso formulazione | 100.052 |