missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)benzeneboronic acid, 97%, Thermo Scientific™

Codice prodotto. 11349106
Cambia vista
Click to view available options
Quantità:
250 mg
1 g
5 g
Dimensione della confezione:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11349106 250 mg 250mg
11329106 1 g 1g
11339106 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11349106 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L20250.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128312-11-8
Formula molecolare C7H9BO2S
Molecular Weight (g/mol) 168.017
Numero MDL MFCD01319105
InChI Key TYVPOLHSKGEXIH-UHFFFAOYSA-N
Sinonimo 3-methylthio phenylboronic acid, 3-methylthio phenyl boronic acid, 3-meththio phenylboronic acid, 3-methylthiophenylboronic acid, 3-methylthio benzeneboronic acid, 3-methylsulfanyl phenylboronic acid, 3-boronothioanisole, 3-methylthiobenzeneboronic acid, 4-methylthiophenyl boronic acid, 3-methylthio phenyl-boronic acid
PubChem CID 2734383
IUPAC Name (3-methylsulfanylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)SC)(O)O

Specifications

Materiale o nome chimico 3-(Methylthio)benzeneboronic acid
Punto di fusione 158°C to 161°C
Quantità 250 mg
Beilstein 7020728
Peso formulazione 168.02
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.