Learn More
3-(Methylthio)butanal, TRC
Product Code. 30477907
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30477907 Supplier Toronto Research Chemicals Supplier No. TRCM3302501G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(Methylthio)butanal
CAS 16630-52-7
Molecular Formula C5H10OS
Quantity 1 g
Synonym 3-(Methylthio)butyraldehyde, 3-Methylsulfanylbutyraldehyde, beta-(Methylthio)butyraldehyde
InChI Formula InChI=1S/C5H10OS/c1-5(7-2)3-4-6/h4-5H,3H2,1-2H3
SMILES CSC(C)CC=O
IUPAC Name 3-methylsulfanylbutanal
Molecular Weight (g/mol) 118.2
Formula Weight 118.0452
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.