Learn More
3-O-Benzyl-1,2,4,6-tetra-O-acetyl-Alpha-D-mannopyranose, TRC
Product Code. 30340845
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340845 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30340845 Supplier Toronto Research Chemicals Supplier No. TRCB296000250MG
 Restricted Item
Spedizione stimata: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-O-Benzyl-1,2,4,6-tetra-O-acetyl-a-D-mannopyranose
CAS 65827-58-9
Molecular Formula C21 H26 O10
Quantity 250 mg
InChI Formula InChI=1S/C21H26O10/c1-12(22)26-11-17-18(28-13(2)23)19(27-10-16-8-6-5-7-9-16)20(29-14(3)24)21(31-17)30-15(4)25/h5-9,17-21H,10-11H2,1-4H3/t17-,18-,19+,20+,21+/m1/s1
SMILES CC(=O)OC[C@H]1O[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H](OCc2ccccc2)[C@@H]1OC(=O)C
IUPAC Name [(2R,3R,4S,5S,6R)-3,5,6-triacetyloxy-4-phenylmethoxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol) 438.43
Formula Weight 438.1526