missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butyldimethylsiloxy-4-methoxybenzaldehyde, TRC

Codice prodotto. 30331141
Cambia vista
Click to view available options
Quantità:
2 g
Dimensione della confezione:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30331141 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30331141 Fornitore Toronto Research Chemicals N. del fornitore TRCB6914852G

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 3-tert-Butyldimethylsiloxy-4-methoxybenzaldehyde
CAS 97315-18-9
Formula molecolare C14H22O3Si
Quantità 2 g
Sinonimo 3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-4-methoxy-benzaldehyde, 3-[(tert-Butyldimethylsilyl)oxy]-4-methoxybenzaldehyde, NSC 620549
InChI Formula InChI=1S/C14H22O3Si/c1-14(2,3)18(5,6)17-13-9-11(10-15)7-8-12(13)16-4/h7-10H,1-6H3
SMILES COc1ccc(C=O)cc1O[Si](C)(C)C(C)(C)C
IUPAC Name 3-[tert-butyl(dimethyl)silyl]oxy-4-methoxybenzaldehyde
Molecular Weight (g/mol) 266.41
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.