Learn More
3-Thiophenecarboxaldehyde, TRC
Product Code. 30455288
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455288 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455288 Supplier Toronto Research Chemicals Supplier No. TRCT3448455G
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Thiophenecarboxaldehyde
CAS 498-62-4
Molecular Formula C5H4OS
Quantity 5 g
Synonym 3-Formylthiophene, 3-Thienaldehyde, 3-Thienylcarboxaldehyde, 3-Thiophenealdehyde, 3-Thiophenecarbaldehyde, NSC 172858, Thiofuran-3-carboxaldehyde, Thiophen-3-aldehyde
InChI Formula InChI=1S/C5H4OS/c6-3-5-1-2-7-4-5/h1-4H
SMILES O=CC1=CSC=C1
IUPAC Name thiophene-3-carbaldehyde
Molecular Weight (g/mol) 112.15
Formula Weight 112