missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzylamine, 96%, Thermo Scientific™

Catalog Number 11404297
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
93071-75-1
C8H8F3NO
191.153
MFCD00061267
TUPUHSXMDIWJQT-UHFFFAOYSA-N
3-trifluoromethoxy benzylamine, 3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy benzyl amine, 3-trifluormethoxybenzylamine, m-trifluoromethoxybenzylamine, 3-trifluoromethoxybenzylamine, benzenemethanamine, 3-trifluoromethoxy, 1-3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy phenyl methylamine
145264
[3-(trifluoromethoxy)phenyl]methanamine
C1=CC(=CC(=C1)OC(F)(F)F)CN
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
93071-75-1
C8H8F3NO
191.153
MFCD00061267
TUPUHSXMDIWJQT-UHFFFAOYSA-N
3-trifluoromethoxy benzylamine, 3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy benzyl amine, 3-trifluormethoxybenzylamine, m-trifluoromethoxybenzylamine, 3-trifluoromethoxybenzylamine, benzenemethanamine, 3-trifluoromethoxy, 1-3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy phenyl methylamine
145264
[3-(trifluoromethoxy)phenyl]methanamine
C1=CC(=CC(=C1)OC(F)(F)F)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-(Trifluoromethoxy)benzylamine
Indice di rifrazione 1.4525
Quantità 5 g
Numero UN UN2735
Beilstein 8763246
Sensibilità Air Sensitive
Peso formulazione 191.15
Percent Purity 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'