Learn More
Rephile™ 30 L Tank Level Sensor
Product Code. 15429659 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15429659 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15429659
  • ONLINE EXCLUSIVE PRICE
    188.51€
    152.60€ / pezzi 1
    Risparmia 35.91€
    19% Sconto
 Restricted Item
Spedizione stimata: 09-12-2026
Add to basket
Add to basket
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
30 L Tank Level Sensor. Reuse the float.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Comparable to FTPF05380
30 L Tank Level Sensor. Comparable to FTPF05380. Reuse the float.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Level Sensor