Learn More
Gilson™ 305 Pump Control Module
Product Code. 16691444 Shop All Gilson Products
Change view
Click to view available options
Max. Flow Rate:
200 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Max. Flow Rate Unit Size
16691444 200 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16691444 Supplier Gilson™ Supplier No. 36100000
  • ONLINE EXCLUSIVE PRICE
    6805.00€
    5521.84€ / pezzi 1
    Risparmia 1283.16€
    18% Sconto
In stock
Together with manometric modules and dynamic mixers, the 305 Pump Control Module provides a cost-effective, dependable pumping solution for analytical to preparative HPLC. Interchangeable pump heads accommodate a variety of liquids at differing viscosities.
  • Wide range of precise and accurate flow rates for isocratic or gradient HPLC
  • Provides high-efficiency mixing for accurate and repeatable gradients over a wide flow rate range
  • Powerful, smooth flow rates from a robust and laboratory-proven pump design
  • Enables simple and easy upgrades from isocratic to binary, tertiary, or quaternary gradients
Specifications
Max. Flow Rate 200 mL/min.
Min. Flow Rate 10 μL/min
Voltage 110/220 V
Product Type HPLC Pump