missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
USP16 Polyclonal antibody specifically detects USP16 in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifica
Specifica
| Antigene | USP16 |
| Applicazioni | ELISA, Western Blot |
| Classificazione | Polyclonal |
| Coniugato | Alexa Fluor 594 |
| Formulazione | 50mM Sodium Borate |
| Alias gene | Deubiquitinating enzyme 16, EC 3.1.2.15, EC 3.4.19.12, human ubiquitin processing protease, EC 3.1.2.1510ubiquitin specific protease 16, ubiquitin carboxyl-terminal hydrolase 16, ubiquitin specific peptidase 16, ubiquitin thioesterase 16, Ubiquitin thiolesterase 16, Ubiquitin-processing protease UBP-M, ubiquitin-specific processing protease 16, Ubiquitin-specific-processing protease 16, UBP-M |
| Specie ospite | Rabbit |
| Immunogeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human USP16 (NP_001001992.1).,, Sequence:, MGKKRTKGKTVPIDDSSETLEPVCRHIRKGLEQGNLKKALVNVEWNICQDCKTDNKVKDKAEEETEEKPSVWLCLKCGHQGCGRNSQEQHALKHYLTPRSEPHCLVLSLDNWSVWCYVCDNEVQYCSSNQLGQVVDYVRKQASITTPKPEKDNGNIELENKKLEKESKNEQEREKKENMAKENPPMNSPCQITVKGLSNLGNTCFFNAVMQNLSQTPVLRELLKEVKMSGTIVKIEPPDLALTEPLEINLEPPGPLTLAMSQFLNEMQETKKGVVTPKEL |
| Metodo di purificazione | Affinity purified |
| Quantità | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?