missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chitinase 3-like 1/YKL-40 Antibody (CL12241) [Janelia Fluor« 646], Novus Biologicals Biologicals™
Sfoglia Tutto Bio Techne ProdottiDescrizione
Chitinase 3-like 1/YKL-40 Monoclonal antibody specifically detects Chitinase 3-like 1/YKL-40 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigene | Chitinase 3-like 1/YKL-40 |
| Applicazioni | Western Blot, Immunohistochemistry (Paraffin) |
| Classificazione | Monoclonal |
| Clone | CL12241 |
| Coniugato | Janelia Fluor 646 |
| Formulazione | 50mM Sodium Borate |
| Alias gene | AW208766, Brp39, Chi3l1, gp39, YKL-40 |
| Specie ospite | Mouse |
| Immunogeno | This antibody was generated using a synthetic peptide of P36222, with the exact immunogen sequence remaining proprietary. |
| Metodo di purificazione | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?