Learn More
Beckman Coulter Life Sciences 3400 Series High-Pressure Diffuser
Product Code. 31777212 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Connection Type:
3/8 in. Barb
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
31777212 3/8 in. Barb Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31777212 Supplier Beckman Coulter Life Sciences Supplier No. 208073212
Spedizione stimata: 27-11-2026
Part of the MET ONE Facility Monitoring System.
  • 3400 Series High Pressure Diffuser for pressurized gas line testing
  • Flow rate 50 L/min., 1.77 CFM
  • 3/8 in. barb connection
Specifications
Product Type High-Pressure Diffuser
Display Type None
Connection Type 3/8 in. Barb
For Use With (Equipment) Beckman Coulter Analyzer