Learn More
3M™ Qualitative Fit-Test Kit Accessories
Codice prodotto. 15749767 Sfoglia Tutto 3M Prodotti
Cambia vista
Click to view available options
Da utilizzare con (applicazione):
Provide secure hood environment
Quantitative Fit Testing
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter
Product Type:
Collar
Fit Test Adapter
Nebulizer
Respirator Test Hood
Dimensione della confezione:
Pezzo
pezzi 2
pezzi 3
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Da utilizzare con (applicazione) Product Type Dimensione della confezione
15749767 Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter Respirator Test Hood pezzi 2
12993731 Provide secure hood environment Collar Pezzo
12973731 Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter Nebulizer pezzi 3
15135157 Quantitative Fit Testing Fit Test Adapter Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 15749767 Fornitore 3M™ N. del fornitore 7000051788
  • ONLINE EXCLUSIVE PRICE
    68.00€
    52.68€ / pezzi 2
    Risparmia 15.32€
    22% Sconto
Spedizione stimata: 04-12-2026
For use with 3M™ Qualitative Fit Test Apparatus
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
TRUSTED_SUSTAINABILITY
Specifica
Product Type Respirator Test Hood
Certificazione/Conformità OSHA
Da utilizzare con (applicazione) Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter
Da utilizzare con (apparecchiatura) Qualitative Fit Test Apparatus FT10 and FT30
Modello FT-14
Quantità 5/Cs.