Learn More
3M™ Versaflo™ M-300 Series PAPR Accessory, Airflow Deflector
Product Code. 15730078 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo M-300 Series Hardhats and M-400 Series Helmets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15730078 Versaflo M-300 Series Hardhats and M-400 Series Helmets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15730078 Supplier 3M™ Supplier No. 7100063940
  • ONLINE EXCLUSIVE PRICE
    59.00€
    40.16€ / pezzi 1
    Risparmia 18.84€
    31% Sconto
Spedizione stimata: 07-01-2027
Airflow deflector is a replacement part for Versaflo™ M-300 Series hardhats and M-400 Series helmets.
  • 3M Catalog# M-316
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Disposable Reusable
Product Type Airflow Deflector
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo M-300 Series Hardhats and M-400 Series Helmets
Model M-316